5-(Boc-amino)-2-bromopyridine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (6-bromopyridin-3-yl)carbamate, 5-Amino-2-bromopyridine, 5-BOC protected
A121583-1G
A121583-5G
103800-090EA
13
USD
103800-090
77312-216
77312-218
5-(Boc-amino)-2-bromopyridine 95%
5-(Boc-amino)-2-bromopyridine
Precaución: for Research Use Only
Formula:
C₁₀H₁₃BrN₂O₂ Boiling Pt: 301 °C (760 mm Hg) Melting Pt: 124…126 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363808 CAS Number: 218594-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...