4-Aminopyrrolidin-2-one, 4-BOC protected 97%
Supplier: Ambeed
|
Warning
|
A235833-10G
A235833-250MG
A235833-1G
103811-038EA
52
USD
103811-038
103850-806
77031-186
77031-188
77089-824
4-Aminopyrrolidin-2-one, 4-BOC protected 97%
4-Aminopyrrolidin-2-one, 4-BOC protected
Precaución: for Research Use Only
Formula:
C₉H₁₆N₂O₃ Boiling Pt: 394.0±31.0 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD17677321 CAS Number: 1245648-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...