tert-Butyl (5-methylpyrazin-2-yl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Boc-amino)-5-methylpyrazine

A325635-250MG A325635-1G
103825-410EA 17 USD
103825-410 103836-356 76788-118 76839-302
tert-Butyl (5-methylpyrazin-2-yl)carbamate 95%
tert-Butyl (5-methylpyrazin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₂
MW: 209.2500 g/mol
Flash Pt: 116.344 °C
Storage Temperature: Freezer
MDL Number: MFCD09907674
CAS Number: 369638-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR