tert-Butyl (5-bromothiazol-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A426906-1G A426906-5G A426906-100G A426906-10G A426906-250MG A426906-25G
103827-478EA 6 USD
103827-478 103838-530 76842-184 76842-186 77168-590 77168-592 77168-594 77307-240
tert-Butyl (5-bromothiazol-2-yl)carbamate 97%
tert-Butyl (5-bromothiazol-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O₂S
MW: 279.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368614
CAS Number: 405939-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR