tert-Butyl (5-bromopentyl)carbamate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(t-Boc-amino)-1-pentyl Bromide
A315550-1G
A315550-100MG
A315550-250MG
A315550-5G
103825-078EA
32
USD
103825-078
76838-832
76875-790
76875-792
77550-880
tert-Butyl (5-bromopentyl)carbamate 95%
tert-Butyl (5-bromopentyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀BrNO₂ MW: 266.18 g/mol Boiling Pt: 324.6 °C (760 mm Hg) Flash Pt: 150.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03425513 CAS Number: 83948-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...