tert-Butyl (5-bromo-4-formylpyridin-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A557965-100MG A557965-250MG
103828-734EA 330 USD
103828-734 77157-236 77157-238
tert-Butyl (5-bromo-4-formylpyridin-2-yl)carbamate 95%
tert-Butyl (5-bromo-4-formylpyridin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18249889
CAS Number: 1260667-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR