tert-Butyl (4-methylpiperidin-4-yl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl N-(4-methylpiperidin-4-yl)carbamate

A277159-100G A277159-10G A277159-250MG A277159-1G A277159-25G A277159-5G
103813-338EA 15 USD
103813-338 103853-274 76787-322 76787-324 76787-326 77093-156 77093-158 77093-160
tert-Butyl (4-methylpiperidin-4-yl)carbamate 97%
tert-Butyl (4-methylpiperidin-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 308 °C (760 mm Hg)
Flash Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08703167
CAS Number: 163271-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR