4-N-Boc-aminophenol 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-hydroxyphenyl)carbamate, 4-(Boc-amino)phenol

A122806-100G A122806-10G A122806-25G A122806-500G A122806-1G A122806-5G
103800-284EA 9 USD
103800-284 76766-642 76766-644 76766-646 76766-648 77312-402 77312-404
4-N-Boc-aminophenol 98%
4-N-Boc-aminophenol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.245 g/mol
Boiling Pt: 288.7 °C (760 mm Hg)
Melting Pt: 143…147 °C
Flash Pt: 128.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00226573
CAS Number: 54840-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR