4-(tert-Butoxycarbonylamino)cyclohexanol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Hydroxycyclohexyl)carbamic acid tert-Butyl ester, tert-Butyl (4-Hydroxycyclohexyl)carbamate, 4-(Boc-amino)cyclohexanol

A124261-100G A124261-10G A124261-1G
103800-508EA 7 USD
103800-508 77009-566 77009-568 77312-642
4-(tert-Butoxycarbonylamino)cyclohexanol 97%
4-(tert-Butoxycarbonylamino)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Boiling Pt: 337.7 °C (760 mm Hg)
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD06658349
CAS Number: 224309-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR