4-(tert-Butoxycarbonylamino)-1-butanol 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-hydroxybutyl)carbamate, N-Boc-4-aminobutanol, 4-(Boc-amino)-1-butanol, N-(4-Hydroxybutyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(4-Hydroxybutyl)carbamate

A427909-100MG A427909-10G A427909-250MG A427909-1G A427909-25G A427909-5G A427909-100G
103827-492EA 9 USD
103827-492 103838-546 76789-188 76789-190 76789-192 76842-208 76842-210 76842-212 77601-838
4-(tert-Butoxycarbonylamino)-1-butanol 97%
4-(tert-Butoxycarbonylamino)-1-butanol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MW: 189.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861334
CAS Number: 75178-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR