tert-Butyl 4-hydroxybutylmethylcarbamate 97%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl (4-hydroxybutyl)(methyl)carbamate

A515011-10G A515011-1G A515011-25G A515011-100MG A515011-250MG
103828-448EA 57 USD
103828-448 103839-552 77169-450 77307-674 77893-046 77893-044 77893-045
tert-Butyl 4-hydroxybutylmethylcarbamate 97%
tert-Butyl 4-hydroxybutylmethylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₃
MW: 203.279 g/mol
Boiling Pt: 284.2±19.0 °C (760 mm Hg)
Flash Pt: 125.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09953073
CAS Number: 99207-32-6
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR