tert-Butyl-4-formylpyridin-3-ylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-formylpyridin-3-yl)carbamate, 3-Aminoisonicotinaldehyde, 3-BOC protected

A184588-100MG A184588-250MG A184588-1G
103815-760EA 52 USD
103815-760 103846-324 76998-800 76998-802 77301-904
tert-Butyl-4-formylpyridin-3-ylcarbamate 97%
tert-Butyl-4-formylpyridin-3-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₃
Boiling Pt: 310 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02683209
CAS Number: 116026-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR