tert-Butyl (4-bromothiophen-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A124765-100MG A124765-10G A124765-250MG A124765-25G A124765-5G A124765-1G
103800-574EA 16 USD
103800-574 76971-788 76971-790 76971-792 76971-794 77298-018 77312-718
tert-Butyl (4-bromothiophen-2-yl)carbamate 95%
tert-Butyl (4-bromothiophen-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO₂S
Boiling Pt: 279 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13195314
CAS Number: 868387-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR