2-(Boc-amino)-4-bromopyridine 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-bromopyridin-2-yl)carbamate

A113985-10G A113985-250MG A113985-25G A113985-1G A113985-5G
103798-328EA 14 USD
103798-328 103824-170 77009-078 77009-080 77009-082 77310-990 77310-992
2-(Boc-amino)-4-bromopyridine 98%
2-(Boc-amino)-4-bromopyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₂
Boiling Pt: 296.7 °C (760 mm Hg)
Melting Pt: 139…140 °C
Flash Pt: 133.239 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09037866
CAS Number: 207799-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR