N-(tert-Butoxycarbonyl)-1,4-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: (4-Aminophenyl)carbamic Acid tert-Butyl Ester, 4-(tert-Butoxycarbonylamino)aniline, N-BOC-p-phenylenediamine, tert-Butyl (4-aminophenyl)carbamate, 4-(Boc-amino)aniline, N-Boc-1,4-phenylenediamine, tert-Butyl (4-Aminophenyl)carbamate

A158358-100G A158358-10G A158358-500G A158358-1G A158358-25G A158358-5G
103805-124EA 6 USD
103805-124 103843-334 76798-874 76798-876 76798-878 77300-324 77319-190 77319-192
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine 98%
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂
Melting Pt: 113 °C
Flash Pt: 132.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00043022
CAS Number: 71026-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR