(4-Amino-phenyl)-methyl-carbamic acid tert-butyl ester 97%

Supplier: Ambeed

Synonyms: tert-Butyl (4-aminophenyl)(methyl)carbamate

A103248-100MG A103248-250MG A103248-1G
103796-514EA 89 USD
103796-514 103822-218 77160-328 77160-330 77309-100
(4-Amino-phenyl)-methyl-carbamic acid tert-butyl ester 97%
(4-Amino-phenyl)-methyl-carbamic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
Boiling Pt: 346.5±25.0 °C (760 mm Hg)
MDL Number: MFCD10693199
CAS Number: 1092522-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR