tert-Butyl (4-aminobut-2-en-1-yl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

A102068-100MG A102068-10G A102068-250MG A102068-25G A102068-1G A102068-5G
103796-278EA 58 USD
103796-278 103821-974 77135-526 77135-528 77135-530 77135-532 77308-856 77308-858
tert-Butyl (4-aminobut-2-en-1-yl)carbamate hydrochloride 95%
tert-Butyl (4-aminobut-2-en-1-yl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O₂
MW: 222.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29918420
CAS Number: 1914155-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR