tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-(Boc-amino)phenylboronic acid Pinacol ester, tert-Butyl [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate, 4-[(tert-Butoxycarbonyl)amino]phenylboronic acid Pinacol ester, 4-Boc-aminophenylboronic acid, pinacol ester, N-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamic acid tert-butyl ester, 4-(Boc-amino)benzeneboronic acid pinacol ester

A167667-1G A167667-5G A167667-10G A167667-250MG A167667-25G A167667-100G
103806-210EA 6 USD
103806-210 103844-468 77081-498 77081-500 77286-662 77286-664 77300-906 77748-115
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97%
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₄
Melting Pt: 167…170 °C
Flash Pt: 208.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02179439
CAS Number: 330793-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR