tert-Butyl 3-oxocyclopentylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-oxocyclopentyl)carbamate

A154379-100MG A154379-10G A154379-250MG A154379-25G A154379-5G A154379-1G
103804-632EA 21 USD
103804-632 103842-820 76973-250 76973-252 76973-254 77300-052 77300-054 77318-534
tert-Butyl 3-oxocyclopentylcarbamate 97%
tert-Butyl 3-oxocyclopentylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MW: 199.247 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09751875
CAS Number: 847416-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR