tert-Butyl (3-oxocyclohexyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Boc-amino)cyclohexanone

A151287-250MG A151287-1G A151287-5G A151287-10G
103804-244EA 9 USD
103804-244 103842-392 76823-830 77317-938 77317-940 77885-243
tert-Butyl (3-oxocyclohexyl)carbamate 95%
tert-Butyl (3-oxocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
Boiling Pt: 335.9 °C (760 mm Hg)
Flash Pt: 157 °C
Storage Temperature: Freezer
MDL Number: MFCD03790905
CAS Number: 885280-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR