tert-Butyl (3-methylpyrrolidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A200929-250MG A200929-100MG
103817-252EA 147 USD
103817-252 103847-888 76825-574 77885-469
tert-Butyl (3-methylpyrrolidin-3-yl)carbamate 95%
tert-Butyl (3-methylpyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Boiling Pt: 289.8±29.0 °C (760 mm Hg)
Flash Pt: 129.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09951966
CAS Number: 147459-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR