tert-Butyl (3-hydroxycyclopentyl)carbamate 97%

Supplier: Ambeed
Warning

A122911-100MG A122911-250MG
103800-306EA 47 USD
103800-306 76781-842 76781-844
tert-Butyl (3-hydroxycyclopentyl)carbamate 97%
tert-Butyl (3-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11975351
CAS Number: 1154870-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR