tert-Butyl (3-hydroxycyclohexyl)carbamate 97%
Supplier: Ambeed
|
Warning
|
A170154-25G
A170154-5G
A170154-10G
A170154-250MG
A170154-1G
103806-452EA
19
USD
103806-452
103844-718
77081-820
77081-822
77163-514
77163-516
77301-024
tert-Butyl (3-hydroxycyclohexyl)carbamate 97%
tert-Butyl (3-hydroxycyclohexyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₁H₂₁NO₃ Flash Pt: 158 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06658389 CAS Number: 610302-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...