4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, 4-(BOC-Amino)-2-fluorophenylboronic acid pinacol ester

A234070-5G A234070-100MG A234070-250MG A234070-1G
103810-924EA 35 USD
103810-924 103850-684 77089-656 77152-954 77152-956 77303-970
4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester 97%
4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BFNO₄
MW: 337.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383441
CAS Number: 1256256-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR