N-Boc-1,3-propanediamine 97%

Supplier: Ambeed
Danger

Synonyms: N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Aminopropyl)carbamate, N-Boc-1,3-propanediamine, tert-Butyl (3-aminopropyl)carbamate, N-1-Boc-1,3-diaminopropane·HCl, N-(tert-Butoxycarbonyl)-1,3-propanediamine, N-Boc-1,3-diaminopropane

A436098-10G A436098-25G A436098-5G A436098-100G
103838-672EA 12 USD
103838-672 76773-996 76842-402 76842-404 77601-882
N-Boc-1,3-propanediamine 97%
N-Boc-1,3-propanediamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
Boiling Pt: 203 °C (760 mm Hg)
Melting Pt: 22 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210021
CAS Number: 75178-96-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR