tert-Butyl (3-acetylphenyl)carbamate 97%

Supplier: Ambeed
Warning

A245236-100MG A245236-250MG A245236-1G A245236-5G
103811-626EA 37 USD
103811-626 103851-464 77031-350 77031-352 77090-704 77090-706
tert-Butyl (3-acetylphenyl)carbamate 97%
tert-Butyl (3-acetylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
Boiling Pt: 310.2 °C (760 mm Hg)
Flash Pt: 141.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09953028
CAS Number: 79537-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR