tert-Butyl (3-(methylamino)propyl)carbamate 97%

Supplier: Ambeed
Warning

A349076-1G A349076-25G A349076-5G A349076-10G A349076-250MG
103826-010EA 9 USD
103826-010 103837-000 76840-084 76840-086 76840-088 77115-104 77115-106
tert-Butyl (3-(methylamino)propyl)carbamate 97%
tert-Butyl (3-(methylamino)propyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₂₀N₂O₂
MW: 188.2673 g/mol
Boiling Pt: 280.3 °C (760 mm Hg)
Flash Pt: 123.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06808586
CAS Number: 442514-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR