tert-Butyl (3-(hydroxymethyl)phenyl)carbamate 97%

Supplier: Ambeed
Warning

A184635-1G A184635-5G A184635-10G A184635-250MG
103815-766EA 55 USD
103815-766 103846-330 77083-946 77083-948 77164-000 77164-002
tert-Butyl (3-(hydroxymethyl)phenyl)carbamate 97%
tert-Butyl (3-(hydroxymethyl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD04974269
CAS Number: 118684-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR