tert-Butyl (3-(dimethylamino)phenyl)carbamate 95%

Supplier: Ambeed
Warning

A137691-100MG A137691-250MG
103802-490EA 50 USD
103802-490 77162-190 77162-192
tert-Butyl (3-(dimethylamino)phenyl)carbamate 95%
tert-Butyl (3-(dimethylamino)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₂
MW: 236.31 g/mol
Boiling Pt: 305.6±25.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD24193076
CAS Number: 1160436-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR