3-(Bromomethyl)aniline, N-BOC protected 97%

Supplier: Ambeed
Danger

A113668-250MG A113668-1G A113668-5G A113668-10G
103798-264EA 25 USD
103798-264 103824-106 76810-064 77310-920 77310-922 77882-940
3-(Bromomethyl)aniline, N-BOC protected 97%
3-(Bromomethyl)aniline, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
Boiling Pt: 309.0±25.0 °C (760 mm Hg)
Melting Pt: 122 °C
Storage Temperature: Freezer
MDL Number: MFCD11974927
CAS Number: 118684-32-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR