t-Boc-N-amido-PEG3-amine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-1-amino-4,7,10-trioxa-13-tridecanamine, tert-Butyl (3-(2-(2-(3-aminopropoxy)ethoxy)ethoxy)propyl)carbamate, N-Boc-C1-PEG3-C3-NH2
A270752-250MG
A270752-1G
A270752-25G
A270752-5G
A270752-100MG
103813-014EA
19
USD
103813-014
103852-920
76692-840
77092-686
77092-688
77092-690
77626-408
t-Boc-N-amido-PEG3-amine 97%
t-Boc-N-amido-PEG3-amine
Precaución: for Research Use Only
Formula:
C₁₅H₃₂N₂O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD04973133 CAS Number: 194920-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...