tert-Butyl (2-oxopyrrolidin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A100880-100MG A100880-10G A100880-250MG A100880-5G
103795-672EA 110 USD
103795-672 76869-122 76869-124 76869-126 76869-128
tert-Butyl (2-oxopyrrolidin-3-yl)carbamate 97%
tert-Butyl (2-oxopyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11007813
CAS Number: 99780-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR