tert-Butyl (2-oxopropyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: (2-Oxopropyl)carbamic acid tert-butyl ester

A311017-1G A311017-5G A311017-100G A311017-100MG A311017-10G A311017-250MG
103814-650EA 8 USD
103814-650 103835-836 76838-624 76838-626 77099-706 77099-708 77099-710 77099-712
tert-Butyl (2-oxopropyl)carbamate 97%
tert-Butyl (2-oxopropyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.2096 g/mol
Boiling Pt: 259.8 °C (760 mm Hg)
Flash Pt: 110.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10698678
CAS Number: 170384-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR