tert-Butyl (2-oxopiperidin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A150095-250MG A150095-1G A150095-5G
103804-124EA 77 USD
103804-124 103842-258 77028-086 77299-782 77299-784
tert-Butyl (2-oxopiperidin-3-yl)carbamate 97%
tert-Butyl (2-oxopiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
Boiling Pt: 405.6 °C (760 mm Hg)
Flash Pt: 199.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13190662
CAS Number: 99780-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR