tert-Butyl (2-oxocyclohexyl)carbamate 95%

Supplier: Ambeed
Warning

A111279-100MG A111279-250MG A111279-5G
103797-810EA 29 USD
103797-810 103823-624 77136-056 77136-058 77310-430
tert-Butyl (2-oxocyclohexyl)carbamate 95%
tert-Butyl (2-oxocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
Boiling Pt: 335.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06411244
CAS Number: 291533-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR