tert-Butyl (2-hydroxyphenyl)carbamate 97%

Supplier: Ambeed
Warning

A107344-10G A107344-250MG A107344-1G A107344-5G
103797-174EA 15 USD
103797-174 103822-942 77008-704 77008-706 77309-790 77309-792
tert-Butyl (2-hydroxyphenyl)carbamate 97%
tert-Butyl (2-hydroxyphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.24 g/mol
Boiling Pt: 265.6 °C (760 mm Hg)
Melting Pt: 140…144 °C
Flash Pt: 114.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06411300
CAS Number: 186663-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR