N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol 97%

Supplier: Ambeed
Warning

A114420-100G A114420-10G A114420-250MG A114420-25G A114420-1G
103798-398EA 7 USD
103798-398 103824-236 76781-282 76781-284 76781-286 76781-288 77297-306 77311-050
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol 97%
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD01883039
CAS Number: 67341-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR