tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Aminoacetohydrazide, 2-BOC protected
A262755-100G
A262755-10G
A262755-250MG
A262755-1G
A262755-25G
A262755-5G
103812-578EA
9
USD
103812-578
103852-474
76771-920
76771-922
76771-924
77092-096
77092-098
77092-100
tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate 95%
tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate
Precaución: for Research Use Only
Formula:
C₇H₁₅N₃O₃ MW: 189.2100 g/mol Melting Pt: 111…115 °C Flash Pt: 187.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01570558 CAS Number: 6926-09-6 UN: 1325 ADR: 4.1 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...