tert-Butyl (2-bromothiophen-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A370769-100MG A370769-250MG A370769-1G
103826-464EA 88 USD
103826-464 103837-480 76876-532 76876-534 77306-816
tert-Butyl (2-bromothiophen-3-yl)carbamate 97%
tert-Butyl (2-bromothiophen-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO₂S
Melting Pt: 65…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01927222
CAS Number: 21483-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR