tert-Butyl (2-bromothiazol-4-yl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A167289-100MG
A167289-250MG
A167289-1G
A167289-5G
103806-158EA
41
USD
103806-158
103844-418
77028-880
77028-882
77081-424
77081-426
tert-Butyl (2-bromothiazol-4-yl)carbamate 98%
tert-Butyl (2-bromothiazol-4-yl)carbamate
Precaución: for Research Use Only
Formula:
C₈H₁₁BrN₂O₂S MW: 279.1541 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12032212 CAS Number: 1245647-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...