4-(Boc-amino)-2-bromopyridine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-bromopyridin-4-yl)carbamate, (2-Bromo-pyridin-4-yl)carbamic acidtert-butyl ester

A193409-1G A193409-10G A193409-250MG A193409-25G
103816-536EA 15 USD
103816-536 103847-152 77085-056 77164-340 77164-342 77302-342
4-(Boc-amino)-2-bromopyridine 97%
4-(Boc-amino)-2-bromopyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₂
MW: 273.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03791266
CAS Number: 433711-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR