tert-Butyl (2-bromophenyl)carbamate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(tert-Butoxycarbonyl)-2-bromoaniline
A114188-25G
A114188-1G
A114188-5G
A114188-10G
103798-362EA
37
USD
103798-362
103824-202
76718-242
77297-292
77297-294
77666-660
tert-Butyl (2-bromophenyl)carbamate 95%
tert-Butyl (2-bromophenyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₁H₁₄BrNO₂ MW: 272.13 g/mol Boiling Pt: 279.2 °C (760 mm Hg) Flash Pt: 122.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD01006614 CAS Number: 78839-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...