tert-Butyl (2-aminophenyl)carbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(tert-Butoxycarbonyl)-1,2-phenylenediamine, tert-Butyl (2-Aminophenyl)carbamate, 2-(tert-Butoxycarbonylamino)aniline, (2-Aminophenyl)carbamic acid tert-Butyl ester, 2-(Boc-amino)aniline, N-Boc-1,2-phenylenediamine
A265306-25G
A265306-5G
A265306-100G
A265306-10G
A265306-250MG
A265306-1G
103812-722EA
6
USD
103812-722
103852-616
77092-292
77092-294
77140-830
77140-832
77140-834
77304-916
tert-Butyl (2-aminophenyl)carbamate 97%
tert-Butyl (2-aminophenyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₁H₁₆N₂O₂ Boiling Pt: 280.6 °C (760 mm Hg) Melting Pt: 109…114 °C Flash Pt: 123.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02169707 CAS Number: 146651-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...