N-Boc-ethylenediamine hydrochloride 95%

Supplier: Ambeed
Warning

A103376-10G A103376-250MG A103376-1G A103376-5G A103376-25G A103376-100G
103796-540EA 6 USD
103796-540 103822-244 76869-254 76869-256 77309-110 77309-112 77570-198 77882-882
N-Boc-ethylenediamine hydrochloride 95%
N-Boc-ethylenediamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₇ClN₂O₂
Boiling Pt: 252.8 °C (760 mm Hg)
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD01076129
CAS Number: 79513-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR