N-Boc-ethylenediamine 97%

Supplier: Ambeed
Danger

Synonyms: N-(tert-Butoxycarbonyl)-1,2-diaminoethane, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl (2-aminoethyl)carbamate, t-boc-N-EDA, tert-Butyl N-(2-Aminoethyl)carbamate, N-BOC-1,2-diaminoethane

A144595-10G A144595-5G A144595-25G
103841-500EA 5 USD
103841-500 76767-958 77299-400 77316-752
N-Boc-ethylenediamine 97%
N-Boc-ethylenediamine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O₂
MW: 160.22 g/mol
Boiling Pt: 252.7 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191871
CAS Number: 57260-73-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR