Boc-glycinamide 97%
Supplier: Ambeed
|
Warning
|
A242927-1G
A242927-5G
A242927-10G
A242927-25G
A242927-100G
103811-494EA
5
USD
103811-494
103851-318
77090-508
77090-510
77165-814
77304-264
77892-435
Boc-glycinamide 97%
Boc-glycinamide
Precaución: for Research Use Only
Formula:
C₇H₁₄N₂O₃ Boiling Pt: 339.7 °C (760 mm Hg) Melting Pt: 141…143 °C Flash Pt: 159.231 °C Storage Temperature: Freezer |
MDL Number:
MFCD08275106 CAS Number: 35150-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...