N-(tert-Butoxycarbonyloxy)succinimide 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2,5-dioxopyrrolidin-1-yl) carbonate

A145479-1G A145479-25G A145479-10G
103803-546EA 5 USD
103803-546 103841-630 77299-456 77316-932 77566-948
N-(tert-Butoxycarbonyloxy)succinimide 98%
N-(tert-Butoxycarbonyloxy)succinimide

Precaución: For research use only.

Formula: C₉H₁₃NO₅
MW: 215.20 g/mol
Melting Pt: 95 °C
Flash Pt: 118.8 °C
Storage Temperature: Freezer
MDL Number: MFCD00037903
CAS Number: 13139-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR