tert-Butyl (2,3-dihydroxypropyl)carbamate 95%

Supplier: Ambeed
Warning

A138890-100G A138890-10G A138890-250MG A138890-500G A138890-25G A138890-1G A138890-5G
103802-674EA 9 USD
103802-674 76688-936 76688-938 76688-940 76688-942 77299-032 77315-734 77315-736
tert-Butyl (2,3-dihydroxypropyl)carbamate 95%
tert-Butyl (2,3-dihydroxypropyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₇NO₄
MW: 191.23 g/mol
Boiling Pt: 361.6 °C (760 mm Hg)
Melting Pt: 60…63 °C
Flash Pt: 172.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01321265
CAS Number: 137618-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR