2-[2-[(tert-Butoxycarbonyl)amino]ethoxy]ethanol 98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl [2-(2-Hydroxyethoxy)ethyl]carbamate, tert-Butyl (2-(2-hydroxyethoxy)ethyl)carbamate, [2-(2-Hydroxyethoxy)ethyl]carbamic acid tert-Butyl ester, 2-[2-(Boc-amino)ethoxy]ethanol

A433873-25G A433873-5G A433873-100G A433873-10G A433873-500G A433873-1G
103827-566EA 5 USD
103827-566 103838-634 76842-346 76842-348 77240-904 77240-906 77240-908 77307-272
2-[2-[(tert-Butoxycarbonyl)amino]ethoxy]ethanol 98%
2-[2-[(tert-Butoxycarbonyl)amino]ethoxy]ethanol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₄
MW: 205.25 g/mol
Boiling Pt: 332.9 °C (760 mm Hg)
Flash Pt: 155.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04038542
CAS Number: 139115-91-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR