tert-Butyl (2-(2-bromoethoxy)ethyl)carbamate 95%

Supplier: Ambeed
Warning

A100475-100MG A100475-250MG A100475-1G A100475-5G A100475-25G A100475-100G
103795-464EA 18 USD
103795-464 103821-158 77148-166 77148-168 77296-706 77296-708 77610-732 77901-682
tert-Butyl (2-(2-bromoethoxy)ethyl)carbamate 95%

Precaución: for Research Use Only

Formula: C₉H₁₈BrNO₃
Storage Temperature: Freezer
MDL Number: MFCD24167474
CAS Number: 164332-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR